missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydrothieno[3,4-b][1,4]dioxine-5-carboxylic acid, ≥97%, Thermo Scientific™

Numéro de catalogue. p-3175043
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
Emballage:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC47801CB 250mg Amber Glass Bottle
CC47801DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC47801CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC47801CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 260063-21-6
Formule moléculaire C7H6O4S
Poids moléculaire (g/mol) 186.181
Numéro MDL MFCD07801102
Clé InChI DCCRIIFBJZOPAV-UHFFFAOYSA-N
Synonyme 2,3-dihydrothieno 3,4-b 1,4 dioxine-5-carboxylic acid, 2,3-dihydro-thieno 3,4-b 1,4 dioxine-5-carboxylic acid, 2h,3h-thieno 3,4-b 1,4 dioxine-5-carboxylic acid, thieno 3,4-b-1,4-dioxin-5-carboxylicacid, 2,3-dihydro, 2h,3h-thiopheno 3,4-e 1,4-dioxane-5-carboxylic acid, 3,4-ethylenedioxythiophene carboxylic acid, thieno 3,4-b-1,4-dioxin-5-carboxylic acid, 2,3-dihydro
PubChem CID 6483880
Nom de l’IUPAC 2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carboxylic acid
SOURIRES C1COC2=C(SC=C2O1)C(=O)O

Spécifications

Nom ou substance chimique 2,3-Dihydrothieno[3,4-b][1,4]dioxine-5-carboxylic acid
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Quantité 250mg
Poids de formule 186.19
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.