missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydrothieno[3,4-b][1,4]dioxine-5-carbonitrile, ≥95%, Thermo Scientific™

Chemical Identifiers

CAS 859851-02-8
Molecular Formula C7H5NO2S
Molecular Weight (g/mol) 167.18
MDL Number MFCD08060540
InChI Key HSQAPDOVCIFQCU-UHFFFAOYSA-N
Synonym 2,3-dihydrothieno 3,4-b 1,4 dioxine-5-carbonitrile, 2h,3h-thieno 3,4-b 1,4 dioxine-5-carbonitrile, thieno 3,4-b-1,4-dioxin-5-carbonitrile,2,3-dihydro, 2h,3h-thiopheno 3,4-e 1,4-dioxane-5-carbonitrile
PubChem CID 7537659
IUPAC Name 2H,3H-thieno[3,4-b][1,4]dioxine-5-carbonitrile
SMILES N#CC1=C2OCCOC2=CS1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC47816CB
View Documents Promotion Details
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC47816DA
View Documents Promotion Details
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

859851-02-8
167.18
HSQAPDOVCIFQCU-UHFFFAOYSA-N
7537659
N#CC1=C2OCCOC2=CS1
C7H5NO2S
MFCD08060540
2,3-dihydrothieno 3,4-b 1,4 dioxine-5-carbonitrile, 2h,3h-thieno 3,4-b 1,4 dioxine-5-carbonitrile, thieno 3,4-b-1,4-dioxin-5-carbonitrile,2,3-dihydro, 2h,3h-thiopheno 3,4-e 1,4-dioxane-5-carbonitrile
2H,3H-thieno[3,4-b][1,4]dioxine-5-carbonitrile

Specifications

859851-02-8
100.0
C7H5NO2S
2,3-dihydrothieno 3,4-b 1,4 dioxine-5-carbonitrile, 2h,3h-thieno 3,4-b 1,4 dioxine-5-carbonitrile, thieno 3,4-b-1,4-dioxin-5-carbonitrile,2,3-dihydro, 2h,3h-thiopheno 3,4-e 1,4-dioxane-5-carbonitrile
N#CC1=C2OCCOC2=CS1
167.18
167.19
2,3-Dihydrothieno[3,4-b][1,4]dioxine-5-carbonitrile
94.5
95%
MFCD08060540
HSQAPDOVCIFQCU-UHFFFAOYSA-N
2H,3H-thieno[3,4-b][1,4]dioxine-5-carbonitrile
7537659
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents