missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydrothieno[3,4-b][1,4]dioxine-5-carbaldehyde, ≥97%, Thermo Scientific™

Numéro de catalogue. CC47804CB
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC47804CB 250mg
CC47804DA 1g
CC47804DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. CC47804CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC47804CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 204905-77-1
Formule moléculaire C7H6O3S
Poids moléculaire (g/mol) 170.182
Numéro MDL MFCD01651766
Clé InChI GNVXYRDVJKJZTO-UHFFFAOYSA-N
Synonyme 2,3-dihydrothieno 3,4-b 1,4 dioxine-5-carbaldehyde, 2h,3h-thieno 3,4-b 1,4 dioxine-5-carbaldehyde, 3,4-ethylenedioxythiophene-2-carbaldehyde, 3,4-ethylenedioxythiophene-2-carboxaldehyde, 2-formyl-3,4-ethylene dioxythiophene, 2h,3h-thiopheno 3,4-e 1,4-dioxane-5-carbaldehyde, 2,3-dihydrothieno 3,4-b-1,4-dioxin-5-carbaldehyde, 2,3-dihydrothieno 3,4-b 1,4 dioxine-5-carboxaldehyde, thieno 3,4-b-1,4-dioxin-5-carboxaldehyde, 2,3-dihydro
PubChem CID 3540090
Nom de l’IUPAC 2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carbaldehyde
SOURIRES C1COC2=C(SC=C2O1)C=O

Spécifications

Nom ou substance chimique 2,3-Dihydrothieno[3,4-b][1,4]dioxine-5-carbaldehyde
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber glass bottle
Quantité 250mg
Poids de formule 170.19
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.