missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydrobenzo[b]furan-7-ylamine, 97%, Thermo Scientific™

Catalog No. CC00914DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC00914DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC00914DA Supplier Thermo Scientific Chemicals Supplier No. CC00914DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13414-56-7
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD11109313
InChI Key UHHZGSLXPQGPJL-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzofuran-7-amine, 2,3-dihydrobenzo b furan-7-ylamine, 2,3-dihydro-benzofuran-7-ylamine, 7-benzofuranamine, 2,3-dihydro, 7-amino-2,3-dihydrobenzofuran, 2,3-dihydro-7-benzofuranamine
PubChem CID 12805950
IUPAC Name 2,3-dihydro-1-benzofuran-7-amine
SMILES NC1=CC=CC2=C1OCC2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 135.17
Percent Purity 97%
Chemical Name or Material 2, 3-Dihydrobenzo[b]furan-7-ylamine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS P Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.