missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-1,4-benzodioxine-5-carbonitrile, 97%, Thermo Scientific™

Numéro de catalogue. p-3484987
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
Emballage:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC01416CB 250mg Amber Glass Bottle
CC01416DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC01416CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC01416CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 148703-14-4
Formule moléculaire C9H7NO2
Poids moléculaire (g/mol) 161.16
Numéro MDL MFCD01116986
Clé InChI WNNQCWLSQDNACP-UHFFFAOYSA-N
Synonyme 2,3-dihydrobenzo b 1,4 dioxine-5-carbonitrile, 1,4-benzodioxin-5-carbonitrile,2,3-dihydro, acmc-1c2ut, 2h,3h-benzo e 1,4-dioxane-5-carbonitrile
PubChem CID 2795043
Nom de l’IUPAC 2,3-dihydro-1,4-benzodioxine-5-carbonitrile
SOURIRES C1COC2=C(C=CC=C2O1)C#N

Spécifications

Nom ou substance chimique 2, 3-Dihydro-1, 4-benzodioxine-5-carbonitrile
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Quantité 250mg
Poids de formule 161.16
Pourcentage de pureté 97%
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED.
IF IN EYES.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.