missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluorobenzoic acid, 98%

Catalog No. AAA1671714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1671714 25 g
AAA1671706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1671714 Supplier Thermo Scientific Chemicals Supplier No. A1671714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4519-39-5 | C7H3F2O2 | 157.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4519-39-5
Molecular Formula C7H3F2O2
Molecular Weight (g/mol) 157.10
MDL Number MFCD00010267
InChI Key JLZVIWSFUPLSOR-UHFFFAOYSA-M
Synonym 2,3-difluoro-benzoic acid, benzoic acid, 2,3-difluoro, difluorobenzoic acid, difluorobenzoic acid;, pubchem1379, 2,3 difluorobenzoic acid, acmc-209k0x, 2,3-difluoro benzoic acid, 2,3-difluorobenzoic acid;, ksc235o6t
PubChem CID 370590
IUPAC Name 2,3-difluorobenzoic acid
SMILES [O-]C(=O)C1=CC=CC(F)=C1F

Specifications

Melting Point 160°C to 164°C
Odor Odorless
Quantity 25 g
Beilstein 2640781
Formula Weight 158.11
Percent Purity 98%
Chemical Name or Material 2,3-Difluorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes

RUO – Research Use Only