missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Difluoro-5-(trifluoromethyl)pyridine 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 104°C |
| Quantity | 5 g |
| Synonym | 2,3-difluoro-5-trifluoromethyl pyridine, 2,3-difluoro-5-trifluoromethyl-pyridine, pyridine,2,3-difluoro-5-trifluoromethyl, pyridine, 2,3-difluoro-5-trifluoromethyl, acmc-20dwi8, 2,3-di-fluoro-5-trifluoromethyl pyridine, alpha,alpha,alpha,5,6-pentafluoro-3-picoline, 2,3-bis fluoranyl-5-trifluoromethyl pyridine |
| Formula Weight | 183.08 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2,3-Difluoro-5-(trifluoromethyl)pyridine |
Safety and Handling
| TSCA | No |