Learn More
Thermo Scientific Chemicals 2',3'-Dideoxyinosine, 98%
CAS: 69655-05-6 | C10H12N4O3 | 236.23 g/mol
Supplier: Thermo Scientific Chemicals J64833MD
| Quantity | 250 mg |
|---|
Description
2?,3?-Dideoxyinosine is a potent anti-retroviral agent. It is most effective in combination therapy for the treatment of HIV and related lymphoma.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2′,3′-Dideoxyinosine is a potent anti-retroviral agent. It is most effective in combination therapy for the treatment of HIV and related lymphoma.
Solubility
Soluble in DMSO or methanol
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Identifiants chimiques
| 69655-05-6 | |
| 236.23 | |
| BXZVVICBKDXVGW-JHPDDGAFNA-N | |
| 50599 | |
| 9-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one |
| C10H12N4O3 | |
| MFCD00077728 | |
| didanosine, 2',3'-dideoxyinosine, dideoxyinosine, videx, videx ec, ddino, inosine, 2',3'-dideoxy, didanosina, didanosinum, ddi | |
| CHEBI:490877 | |
| OC[C@@H]1CC[C@@H](O1)N1C=NC2=C1N=CNC2=O |
Spécifications
| 2',3'-Dideoxyinosine | |
| White | |
| 250 mg | |
| 3619529 | |
| didanosine, 2',3'-dideoxyinosine, dideoxyinosine, videx, videx ec, ddino, inosine, 2',3'-dideoxy, didanosina, didanosinum, ddi | |
| BXZVVICBKDXVGW-JHPDDGAFNA-N | |
| 9-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one | |
| 50599 | |
| 236.23 | |
| Powder |
| 69655-05-6 | |
| C10H12N4O3 | |
| MFCD00077728 | |
| 143098 | |
| Soluble in DMSO or methanol | |
| OC[C@@H]1CC[C@@H](O1)N1C=NC2=C1N=CNC2=O | |
| 236.23 | |
| CHEBI:490877 | |
| 98% |
Sécurité et manipulation
RTECSNumber : NM7460700
TSCA : No
Recommended Storage : Store at -20°C