missing translation for 'onlineSavingsMsg'
Learn More

2',3'-Dideoxyadenosine 98.0+%, TCI America™

Catalog No. D3065100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3065100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D3065100MG Supplier TCI America Supplier No. D3065100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4097-22-7
Molecular Formula C10H13N5O2
Molecular Weight (g/mol) 235.25
MDL Number MFCD00010534
InChI Key WVXRAFOPTSTNLL-NKWVEPMBSA-N
Synonym 2',3'-dideoxyadenosine, dideoxyadenosine, adenosine, 2',3'-dideoxy, ddado, 2s,5r-5-6-amino-9h-purin-9-yl tetrahydrofuran-2-yl methanol, ccris 428, unii-4q86ah641a, d2a, dsstox_cid_3771, dsstox_rid_77189
PubChem CID 20039
IUPAC Name [(2S,5R)-5-(6-amino-9H-purin-9-yl)oxolan-2-yl]methanol
SMILES NC1=C2N=CN([C@H]3CC[C@@H](CO)O3)C2=NC=N1

Specifications

Melting Point 188°C
Color White
Quantity 100 mg
Formula Weight 235.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′,3′-Dideoxyadenosine
RTECSNumber AU7358900
TSCA No