missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dicyanopyrazine 99.0+%, TCI America™

Catalog No. D17515G
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17515G 5 g
D175125G 25 g
D1751100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D17515G Supplier TCI America Supplier No. D17515G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13481-25-9
Molecular Formula C6H2N4
Molecular Weight (g/mol) 130.11
MDL Number MFCD00059771
InChI Key OTVZGAXESBAAQQ-UHFFFAOYSA-N
Synonym Pyrazine-2,3-dicarbonitrile
PubChem CID 300514
IUPAC Name pyrazine-2,3-dicarbonitrile
SMILES C1=CN=C(C(=N1)C#N)C#N

Specifications

Melting Point 133°C
Quantity 5 g
Formula Weight 130.11
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dicyanopyrazine
EINECSNumber (9)-0533&(9)-1651
TSCA No