missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloropyridine-4-boronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. D50811G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D50811G Supplier TCI America Supplier No. D50811G
Only null left

Specifications

Chemical Name or Material 2,3-Dichloropyridine-4-boronic Acid (contains varying amounts of Anhydride)
CAS 951677-39-7
Melting Point 141°C
Color White
Molecular Formula C5H4BCl2NO2
Quantity 1 g
MDL Number MFCD04038756
Synonym 2,3-dichloropyridine-4-boronic acid, 2,3-dichloro-4-pyridineboronic acid, 2,3-dichloropyridin-4-yl boronic acid, 2,3-dichloropyridine-4-boronicacid, 2,3-dichloro-4-pyridineboronicacid, pubchem7941, 2,3-dichloropyridin-4-yl-4-boronic acid, acmc-209rv3, 4-borono-2,3-dichloropyridine, 2,3-dichloro-4-pyridylboronic acid
InChI Key ABRTZUPJNVJMHP-UHFFFAOYSA-N
SMILES OB(O)C1=C(Cl)C(Cl)=NC=C1
IUPAC Name (2,3-dichloropyridin-4-yl)boronic acid
Molecular Weight (g/mol) 191.80
PubChem CID 3478955
Formula Weight 191.80
Physical Form Crystalline Powder
Show More Show Less