missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dichlorophenyl Isothiocyanate 98.0+%, TCI America™
$80.85 - $356.16
Chemical Identifiers
| CAS | 6590-97-2 |
|---|---|
| Molecular Formula | C7H3Cl2NS |
| Molecular Weight (g/mol) | 204.07 |
| MDL Number | MFCD00041048 |
| InChI Key | RQZIODPVCCTBAQ-UHFFFAOYSA-N |
| Synonym | 2,3-dichlorophenyl isothiocyanate, 2,3-dichlorophenylisothiocyanate, benzene,1,2-dichloro-3-isothiocyanato, benzene, 1,2-dichloro-3-isothiocyanato, dichlorophenylisothiocyanate, dichlorophenyl isothiocyanate, acmc-1b4g1, 2,3-dichlorophenyl-isothiocyanate, 1,2-dichloro-3-isothiocyanato-benzene, 2,3-dichloro-1-isothiocyanato-benzene |
| PubChem CID | 2796281 |
| IUPAC Name | 1,2-dichloro-3-isothiocyanatobenzene |
| SMILES | ClC1=CC=CC(N=C=S)=C1Cl |
Chemical Identifiers
| 6590-97-2 | |
| 204.07 | |
| RQZIODPVCCTBAQ-UHFFFAOYSA-N | |
| 2796281 | |
| ClC1=CC=CC(N=C=S)=C1Cl |
| C7H3Cl2NS | |
| MFCD00041048 | |
| 2,3-dichlorophenyl isothiocyanate, 2,3-dichlorophenylisothiocyanate, benzene,1,2-dichloro-3-isothiocyanato, benzene, 1,2-dichloro-3-isothiocyanato, dichlorophenylisothiocyanate, dichlorophenyl isothiocyanate, acmc-1b4g1, 2,3-dichlorophenyl-isothiocyanate, 1,2-dichloro-3-isothiocyanato-benzene, 2,3-dichloro-1-isothiocyanato-benzene | |
| 1,2-dichloro-3-isothiocyanatobenzene |
Specifications
| 6590-97-2 | |
| 258°C | |
| MFCD00041048 | |
| 1760 | |
| RQZIODPVCCTBAQ-UHFFFAOYSA-N | |
| 1,2-dichloro-3-isothiocyanatobenzene | |
| 2796281 | |
| ≥98.0% (GC) | |
| 2,3-Dichlorophenyl Isothiocyanate |
| Yellow | |
| C7H3Cl2NS | |
| 5 g | |
| 2,3-dichlorophenyl isothiocyanate, 2,3-dichlorophenylisothiocyanate, benzene,1,2-dichloro-3-isothiocyanato, benzene, 1,2-dichloro-3-isothiocyanato, dichlorophenylisothiocyanate, dichlorophenyl isothiocyanate, acmc-1b4g1, 2,3-dichlorophenyl-isothiocyanate, 1,2-dichloro-3-isothiocyanato-benzene, 2,3-dichloro-1-isothiocyanato-benzene | |
| ClC1=CC=CC(N=C=S)=C1Cl | |
| 204.07 | |
| 204.07 | |
| Liquid |
Safety and Handling
TSCA : No