missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dichlorophenyl Isothiocyanate 98.0+%, TCI America™
Supplier: TCI America D492325G
Specifications
| 2,3-Dichlorophenyl Isothiocyanate | |
| Yellow | |
| C7H3Cl2NS | |
| 25 g | |
| 2,3-dichlorophenyl isothiocyanate, 2,3-dichlorophenylisothiocyanate, benzene,1,2-dichloro-3-isothiocyanato, benzene, 1,2-dichloro-3-isothiocyanato, dichlorophenylisothiocyanate, dichlorophenyl isothiocyanate, acmc-1b4g1, 2,3-dichlorophenyl-isothiocyanate, 1,2-dichloro-3-isothiocyanato-benzene, 2,3-dichloro-1-isothiocyanato-benzene | |
| ClC1=CC=CC(N=C=S)=C1Cl | |
| 204.07 | |
| 204.07 | |
| Liquid |
| 6590-97-2 | |
| 258°C | |
| MFCD00041048 | |
| 1760 | |
| RQZIODPVCCTBAQ-UHFFFAOYSA-N | |
| 1,2-dichloro-3-isothiocyanatobenzene | |
| 2796281 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 6590-97-2 | |
| 204.07 | |
| RQZIODPVCCTBAQ-UHFFFAOYSA-N | |
| 2796281 | |
| ClC1=CC=CC(N=C=S)=C1Cl |
| C7H3Cl2NS | |
| MFCD00041048 | |
| 2,3-dichlorophenyl isothiocyanate, 2,3-dichlorophenylisothiocyanate, benzene,1,2-dichloro-3-isothiocyanato, benzene, 1,2-dichloro-3-isothiocyanato, dichlorophenylisothiocyanate, dichlorophenyl isothiocyanate, acmc-1b4g1, 2,3-dichlorophenyl-isothiocyanate, 1,2-dichloro-3-isothiocyanato-benzene, 2,3-dichloro-1-isothiocyanato-benzene | |
| 1,2-dichloro-3-isothiocyanatobenzene |
Safety and Handling
TSCA : No