accessibility menu, dialog, popup

UserName

CAS RN 6590-97-2

IUPAC Name: 1,2-dichloro-3-isothiocyanatobenzene
Synonyms: 1,2-dichloro-3-isothiocyanatobenzene
Molecular Weight (g/mol): 204.07
Molecular Formula: C7H3Cl2NS
InChi Key: RQZIODPVCCTBAQ-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(N=C=S)=C1Cl
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)