missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzylamine 98.0+%, TCI America™

Catalog No. D29245G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D29245G Supplier TCI America Supplier No. D29245G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 39226-95-4
Molecular Formula C7H7Cl2N
Molecular Weight (g/mol) 176.04
MDL Number MFCD00047927
InChI Key JHBVZGONNIVXFJ-UHFFFAOYSA-N
Synonym 2,3-dichlorobenzylamine, 2,3-dichlorophenyl methanamine, 2,3-dichloro-benzylamine, 2,3-dichlorobenzyl amine, chembl13165, 1-2,3-dichlorophenyl methanamine, benzenemethanamine, 2,3-dichloro, 2,3-dichlorophenyl methylamine, pubchem16713, acmc-1ae0r
PubChem CID 587625
IUPAC Name (2,3-dichlorophenyl)methanamine
SMILES C1=CC(=C(C(=C1)Cl)Cl)CN

Specifications

Color Yellow
Boiling Point 140°C
Quantity 5 g
UN Number 2735
Formula Weight 176.04
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,3-Dichlorobenzylamine