missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. D35685G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D35685G Supplier TCI America Supplier No. D35685G

Chemical Identifiers

CAS 82417-45-6
Molecular Formula C6H3Cl3O2S
Molecular Weight (g/mol) 245.498
MDL Number MFCD00051844
InChI Key PQODWTNHDKDHIW-UHFFFAOYSA-N
Synonym 2,3-dichlorobenzene-1-sulfonyl chloride, 2,3-dichlorobenzenesulphonyl chloride, 2,3-dichloro-benzenesulfonyl chloride, 2,3-dichlorobenzenesulphonylchloride, 2,3-dichlorophenylsulfonyl chloride, benzenesulfonyl chloride, 2,3-dichloro-9ci, 2,3-dichlorobenzenesulfonylchloride, pubchem5114, buttpark 9357-48, acmc-1bl9r
PubChem CID 2734222
IUPAC Name 2,3-dichlorobenzenesulfonyl chloride
SMILES C1=CC(=C(C(=C1)Cl)Cl)S(=O)(=O)Cl

Specifications

Melting Point 62°C
Color White-Yellow
Boiling Point 142°C
Quantity 5 g
UN Number 3261
Formula Weight 245.50
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dichlorobenzenesulfonyl Chloride