missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloroaniline, 99%

Catalog No. AAA1152436
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1152436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1152436 Supplier Thermo Scientific Chemicals Supplier No. A1152436
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 608-27-5 | C6H5Cl2N | 162.013 g/mol

2,3-Dichloroaniline is used in dyestuffs, pigments and optical brighteners. It is used as flame retardants for polymers. It is used in the manufacturing of growth regulators, polymer auxiliaries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dichloroaniline is used in dyestuffs, pigments and optical brighteners. It is used as flame retardants for polymers. It is used in the manufacturing of growth regulators, polymer auxiliaries.

Solubility
Soluble in ether, alcohol and acetone. Slightly soluble in benzene. Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.

Chemical Identifiers

CAS 608-27-5
Molecular Formula C6H5Cl2N
Molecular Weight (g/mol) 162.013
MDL Number MFCD00007657
InChI Key BRPSAOUFIJSKOT-UHFFFAOYSA-N
Synonym benzenamine, 2,3-dichloro, 2,3-dichlorobenzenamine, aniline, 2,3-dichloro, unii-2bl4f1dxvn, 2,3-dichloranilin, 2,3 dichloraniline, 2,3-dichloro-phenylamine, 2bl4f1dxvn, aniline, 2,3-dichloro-7ci,8ci, 2,3-dichlorophenylamine
PubChem CID 11844
ChEBI CHEBI:46636
IUPAC Name 2,3-dichloroaniline
SMILES C1=CC(=C(C(=C1)Cl)Cl)N

Specifications

Melting Point 20°C to 24°C
Density 1.37
Boiling Point 250°C to 252°C
Flash Point 115°C (239°F)
Refractive Index 1.606
Quantity 500 g
UN Number UN3442
Beilstein 472027
Solubility Information Soluble in ether,alcohol and acetone. Slightly soluble in benzene. Insoluble in water.
Formula Weight 162.02
Percent Purity 99%
Chemical Name or Material 2,3-Dichloroaniline
Show More Show Less
Hazard Category H301+H311+H331-H315-H319-H500
Hazard Statement GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DICHLOROANILINES, SOLID
EINECSNumber 210-157-9
RTECSNumber CX9862625
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only