missing translation for 'onlineSavingsMsg'
Learn More

2',3'-Dichloroacetophenone, 98%

Numéro de catalogue. AAA1266703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAA1266703 1 g
AAA1266706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAA1266703 Fournisseur Thermo Scientific Chemicals Code fournisseur A1266703
Il en reste null

CAS: 56041-57-7 | C8H6Cl2O | 189.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 56041-57-7
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.04
MDL Number MFCD00052988
InChI Key KMABBMYSEVZARZ-UHFFFAOYSA-N
Synonym 1-2,3-dichlorophenyl ethanone, 2',3'-dichloroacetophenone, 2,3-dichloroacetophenone, 1-2,3-dichlorophenyl ethan-1-one, ethanone, 1-2,3-dichlorophenyl, 1-acetyl-2,3-dichlorobenzene, pubchem3368, 1-2,3-dichlorophenyl-1-ethanone, 1-2,3-bis chloranyl phenyl ethanone
PubChem CID 91986
IUPAC Name 1-(2,3-dichlorophenyl)ethanone
SMILES CC(=O)C1=CC=CC(Cl)=C1Cl

Spécifications

Density 1.293
Boiling Point 106°C to 108°C (2 mmHg)
Refractive Index 1.56
Quantity 1 g
Beilstein 2438815
Formula Weight 189.04
Percent Purity 98%
Chemical Name or Material 2',3'-Dichloroacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 259-954-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only