missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-1-propene 97.0+%, TCI America™

Catalog No. D040625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D040625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D040625G Supplier TCI America Supplier No. D040625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 78-88-6
Color Yellow
Boiling Point 94°C
Molecular Formula C3H4Cl2
Quantity 25 g
MDL Number MFCD00000943
UN Number 2047
Synonym 2,3-dichloropropene, 2,3-dichloro-1-propene, 2-chloroallyl chloride, 2,3-dichloropropylene, 1-propene, 2,3-dichloro, propene, 2,3-dichloro, 1,2-dichloro-2-propene, propylene, 2,3-dichloro, ccris 956, unii-370tnk5i41
InChI Key FALCMQXTWHPRIH-UHFFFAOYSA-N
SMILES C=C(CCl)Cl
IUPAC Name 2,3-dichloroprop-1-ene
Molecular Weight (g/mol) 110.965
PubChem CID 6565
Formula Weight 110.97
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Dichloro-1-propene
Show More Show Less
RTECSNumber UC8400000
TSCA Yes
Recommended Storage Refrigerator