missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromo-5-methylpyridine 98.0+%, TCI America™

Catalog No. D51005G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D51005G Supplier TCI America Supplier No. D51005G

Specifications

CAS 29232-39-1
Melting Point 55°C
Boiling Point 271°C
Molecular Formula C6H5Br2N
Quantity 5 g
MDL Number MFCD04112574
Synonym 2,3-dibromo-5-picoline, 5,6-dibromo-3-picoline, pyridine, 2,3-dibromo-5-methyl, pubchem5399, acmc-209h7j, ksc562s8b, 2,3 dibromo-5-methyl pyridine, 2,3-dibromo-5-methyl-pyridine, 5,6-dibromo-m-picoline, pyridine,2,3-dibromo-5-methyl
InChI Key BZUZAZMAGVFIBS-UHFFFAOYSA-N
SMILES CC1=CC(=C(N=C1)Br)Br
IUPAC Name 2,3-dibromo-5-methylpyridine
Molecular Weight (g/mol) 250.921
PubChem CID 285435
Formula Weight 250.92
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dibromo-5-methylpyridine
Show More Show Less