missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromo-4-hydroxy-5-methoxybenzaldehyde, 98%

Catalog No. AAB2114014
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2114014 25 g
AAB2114003 1 g
AAB2114006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2114014 Supplier Thermo Scientific Chemicals Supplier No. B2114014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2973-75-3 | C8H6Br2O3 | 309.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2973-75-3
Molecular Formula C8H6Br2O3
Molecular Weight (g/mol) 309.94
MDL Number MFCD00016978
InChI Key WKLKGSHBXNPUDU-UHFFFAOYSA-N
Synonym 5,6-dibromovanillin, acmc-1chm2, 2,3-dibromo-5-methoxy-4-hydroxybenzaldehyde, 2,3-bis bromanyl-5-methoxy-4-oxidanyl-benzaldehyde
PubChem CID 520452
IUPAC Name 2,3-dibromo-4-hydroxy-5-methoxybenzaldehyde
SMILES COC1=CC(C=O)=C(Br)C(Br)=C1O

Specifications

Melting Point 213°C to 217°C
Quantity 25 g
Beilstein 1968576
Sensitivity Air Sensitive
Formula Weight 309.94
Percent Purity 98%
Chemical Name or Material 2,3-Dibromo-4-hydroxy-5-methoxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only