missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromo-3-phenylpropionic Acid 98.0+%, TCI America™

Catalog No. D264725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D264725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D264725G Supplier TCI America Supplier No. D264725G
Only null left

Chemical Identifiers

CAS 6286-30-2
Molecular Formula C9H8Br2O2
Molecular Weight (g/mol) 307.97
MDL Number MFCD00004208
InChI Key FXJWTHBNVZNQQP-UHFFFAOYNA-N
Synonym alpha,beta-Dibromohydrocinnamic Acid
PubChem CID 22705
IUPAC Name 2,3-dibromo-3-phenylpropanoic acid
SMILES OC(=O)C(Br)C(Br)C1=CC=CC=C1

Specifications

Melting Point 199°C
Color White-Yellow
Quantity 25 g
Formula Weight 307.97
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dibromo-3-phenylpropionic Acid
RTECSNumber MW5135000
TSCA No