missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Chlorophenoxy)propionic Acid 98.0+%, TCI America™

Catalog No. C109425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C109425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C109425G Supplier TCI America Supplier No. C109425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 101-10-0
Molecular Formula C9H9ClO3
Molecular Weight (g/mol) 200.618
MDL Number MFCD00009723
InChI Key YNTJKQDWYXUTLZ-UHFFFAOYSA-N
Synonym 2-3-chlorophenoxy propionic acid, cloprop, 2-3-chlorophenoxy propanoic acid, metachlorphenprop, fruitone cpa, propanoic acid, 2-3-chlorophenoxy, 2-3-chlorophenoxy-propionic acid, amchem 3-cp, caswell no. 206, cloprop iso:prop
PubChem CID 7542
IUPAC Name 2-(3-chlorophenoxy)propanoic acid
SMILES CC(C(=O)O)OC1=CC(=CC=C1)Cl

Specifications

Melting Point 114°C
Quantity 25 g
Formula Weight 200.62
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(3-Chlorophenoxy)propionic Acid
RTECSNumber UE9285000
TSCA Yes