missing translation for 'onlineSavingsMsg'
Learn More

2-[3-(Chloromethyl)phenyl]pyrimidine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC56324DA

Quantity 1g
Packaging Amber Glass Bottle

 Afficher plus de versions de ce produit

Numéro de catalogue. CC56324DA

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.


Description

Description

Spécifications

Identifiants chimiques

898289-51-5
204.657
AUWJTKGKFPDHDU-UHFFFAOYSA-N
24229567
C1=CC(=CC(=C1)CCl)C2=NC=CC=N2
C11H9ClN2
MFCD09064978
2-3-chloromethyl phenyl pyrimidine, 3-pyrimidin-2-ylbenzyl chloride, pyrimidine, 2-3-chloromethyl phenyl, pyrimidine, 2-3-chloromethyl phenyl-9ci
2-[3-(chloromethyl)phenyl]pyrimidine

Spécifications

2-[3-(Chloromethyl)phenyl]pyrimidine
96.5
97%
C11H9ClN2
MFCD09064978
AUWJTKGKFPDHDU-UHFFFAOYSA-N
2-[3-(chloromethyl)phenyl]pyrimidine
24229567
97%
898289-51-5
100.0
Amber Glass Bottle
1g
2-3-chloromethyl phenyl pyrimidine, 3-pyrimidin-2-ylbenzyl chloride, pyrimidine, 2-3-chloromethyl phenyl, pyrimidine, 2-3-chloromethyl phenyl-9ci
C1=CC(=CC(=C1)CCl)C2=NC=CC=N2
204.657
204.66
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

FDS
Documents

Documents

Promotions

Promotions