missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Bromophenyl)-4,6-diphenyl-1,3,5-triazine 97.0+%, TCI America™

Catalog No. B48945G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48945G 5 g
B48941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48945G Supplier TCI America Supplier No. B48945G
Only null left

Chemical Identifiers

CAS 864377-31-1
Molecular Formula C21H14BrN3
Molecular Weight (g/mol) 388.27
MDL Number MFCD22200060
InChI Key HNZUKQQNZRMNGS-UHFFFAOYSA-N
PubChem CID 58943302
IUPAC Name 2-(3-bromophenyl)-4,6-diphenyl-1,3,5-triazine
SMILES BrC1=CC=CC(=C1)C1=NC(=NC(=N1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 176°C
Color White
Quantity 5 g
Formula Weight 388.27
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-(3-Bromophenyl)-4,6-diphenyl-1,3,5-triazine