missing translation for 'onlineSavingsMsg'
Learn More

2-[3-(Bromomethyl)phenyl]thiophene, ≥97%, Thermo Scientific™

Numéro de catalogue. p-3174844
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC44508CB 250mg
CC44508DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC44508CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC44508CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 85553-44-2
Formule moléculaire C11H9BrS
Poids moléculaire (g/mol) 253.157
Numéro MDL MFCD08435867
Clé InChI JQQDEBDYUAKNGD-UHFFFAOYSA-N
Synonyme 2-3-bromomethyl phenyl thiophene, 3-2-thienyl benzyl bromide, 2-3-bromomethyl phenyl-thiophene
PubChem CID 13150400
Nom de l’IUPAC 2-[3-(bromomethyl)phenyl]thiophene
SOURIRES C1=CC(=CC(=C1)CBr)C2=CC=CS2

Spécifications

Nom ou substance chimique 2-[3-(Bromomethyl)phenyl]thiophene
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Quantité 250mg
Poids de formule 253.16
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.