missing translation for 'onlineSavingsMsg'
Learn More

2,3-Bis(2,6-diisopropylphenylimino)butane 98.0+%, TCI America™

Catalog No. B5606200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5606200MG 200 mg
B56061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. B5606200MG missing translation for 'mfr' TCI America missing translation for 'supplierNo' B5606200MG
Only null left

Chemical Identifiers

CAS 74663-77-7
Molecular Formula C28H40N2
Molecular Weight (g/mol) 404.642
MDL Number MFCD01862435
InChI Key YUFQUBWPYIPRHZ-UHFFFAOYSA-N
Synonym N,N′-Bis(2,6-diisopropylphenyl)butane-2,3-diimine
PubChem CID 608871
IUPAC Name 2-N,3-N-bis[2,6-di(propan-2-yl)phenyl]butane-2,3-diimine
SMILES CC(C)C1=C(C(=CC=C1)C(C)C)N=C(C)C(=NC2=C(C=CC=C2C(C)C)C(C)C)C

Specifications

Melting Point 101°C
Quantity 200 mg
Formula Weight 404.64
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Bis(2,6-diisopropylphenylimino)butane