missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Bis(2,4,5-trimethyl-3-thienyl)maleimide 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 181°C |
| Quantity | 1 g |
| Formula Weight | 345.48 |
| Percent Purity | ≥97.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,3-Bis(2,4,5-trimethyl-3-thienyl)maleimide |
Safety and Handling
| TSCA | No |