missing translation for 'onlineSavingsMsg'
Learn More

2,3,6,7,10,11-Hexakis[(n-octyl)oxy]triphenylene 98.0+%, TCI America™

Catalog No. p-7131050
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1450200MG 200 mg
H14501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H1450200MG Supplier TCI America Supplier No. H1450200MG
Only null left

Chemical Identifiers

CAS 70351-87-0
Molecular Formula C66H108O6
Molecular Weight (g/mol) 997.58
MDL Number MFCD20528916
InChI Key IUFDADHEWRYRSP-UHFFFAOYSA-N
PubChem CID 15219108
IUPAC Name 2,3,6,7,10,11-hexakis(octyloxy)triphenylene
SMILES CCCCCCCCOC1=C(OCCCCCCCC)C=C2C(=C1)C1=CC(OCCCCCCCC)=C(OCCCCCCCC)C=C1C1=CC(OCCCCCCCC)=C(OCCCCCCCC)C=C21

Specifications

Color Yellow
Quantity 200 mg
Formula Weight 997.58
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,6,7,10,11-Hexakis[(n-octyl)oxy]triphenylene