missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Triphenyltetrazolium Chloride 98.0+%, TCI America™

Catalog No. T05201G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T05201G 1 g
T052025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T05201G Supplier TCI America Supplier No. T05201G

Chemical Identifiers

CAS 298-96-4
Molecular Formula C19H15ClN4
Molecular Weight (g/mol) 334.81
MDL Number MFCD00011963
InChI Key PKDBCJSWQUOKDO-UHFFFAOYSA-M
Synonym 2,3,5-triphenyltetrazolium chloride, tetrazolium red, uroscreen, red tetrazolium, urocheck, vitastain, tetrazolium chloride, triphenyltetrazolium chloride, 2,3,5-triphenyl-2h-tetrazolium chloride, tetrazolium chloride
PubChem CID 9283
ChEBI CHEBI:78019
IUPAC Name triphenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
SMILES [Cl-].C1=CC=C(C=C1)N1N=C(N=[N+]1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 334.81
Percent Purity ≥98.0% (T)
Physical Form Powder
Chemical Name or Material 2,3,5-Triphenyltetrazolium Chloride
EINECSNumber (5)-3612
RTECSNumber XF8100000
TSCA Yes