missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(3,5-Di-tert-butyl-2-hydroxyphenyl)-5-chlorobenzotriazole 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 155°C |
| Color | Yellow |
| Quantity | 25 g |
| Formula Weight | 357.88 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-(3,5-Di-tert-butyl-2-hydroxyphenyl)-5-chlorobenzotriazole |
Safety and Handling
| EINECSNumber | (5)-3581&(5)-3605 |
| RTECSNumber | SK3425000 |
| TSCA | Yes |