missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrafluoro-1,4-benzenedimethanol 97.0+%, TCI America™

Catalog No. T27325G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27325G 5 g
T273225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27325G Supplier TCI America Supplier No. T27325G
Only null left

Chemical Identifiers

CAS 92339-07-6
Molecular Formula C8H6F4O2
Molecular Weight (g/mol) 210.128
MDL Number MFCD00229147
InChI Key SDHKGYDQOGCLQM-UHFFFAOYSA-N
PubChem CID 4098835
IUPAC Name [2,3,5,6-tetrafluoro-4-(hydroxymethyl)phenyl]methanol
SMILES C(C1=C(C(=C(C(=C1F)F)CO)F)F)O

Specifications

Melting Point 129°C
Color White-Yellow
Quantity 5 g
Synonym 2,3,5,6-Tetrafluoro-1,4-bis(hydroxymethyl)benzene, 2,3,5,6-Tetrafluoro-alpha,alpha′C-dihydroxy-p-xylene, 2,3,5,6-Tetrafluoro-p-xylene-alpha,alpha′C-diol
Formula Weight 210.13
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,5,6-Tetrafluoro-1,4-benzenedimethanol