missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,5,6-Tetrabromo-p-cresol 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 198°C |
| Color | White |
| Quantity | 5 g |
| Synonym | 2,3,5,6-tetrabromo-p-cresol, tetrabromo-p-cresol, 2,3,5,6-tetrabromo-4-methyl-phenol, phenol,2,3,5,6-tetrabromo-4-methyl, phenol, 2,3,5,6-tetrabromo-4-methyl, tetrabrom-p-kresol, acmc-1cswl, 2,3,5,6-tetrabromo-4-methylphe, 4-methyl-2,3,5,6-tetrabromophenol, # |
| Formula Weight | 423.72 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,3,5,6-Tetrabromo-p-cresol |
Safety and Handling
| TSCA | No |