missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trihydroxybenzaldehyde, 98%

Catalog No. AAA1374506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1374506 5 g
AAA1374514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1374506 Supplier Thermo Scientific Chemicals Supplier No. A1374506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2144-08-3 | C8H9BO2 | 147.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2144-08-3

Specifications

Melting Point 159°C to 163°C
Molecular Formula C8H9BO2
MDL Number MFCD00239441
Quantity 5 g
Beilstein 2328658
Sensitivity Air Sensitive
Synonym 4-vinylphenylboronic acid, 4-vinylbenzeneboronic acid, 4-vinylphenyl boronic acid, 4-ethenylphenyl boronic acid, 4-vinylboronic acid, p-styrylboronic acid, p-vinylphenylboronic acid, styrene-4-boronic acid, boronic acid, 4-ethenylphenyl, 4-styreneboronic acid
InChI Key QWMJEUJXWVZSAG-UHFFFAOYSA-N
SMILES OB(O)C1=CC=C(C=C)C=C1
IUPAC Name (4-ethenylphenyl)boronic acid
Molecular Weight (g/mol) 147.97
PubChem CID 2734393
Formula Weight 154.12
Percent Purity 98%
Chemical Name or Material 2,3,4-Trihydroxybenzaldehyde
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 218-404-2
RTECSNumber CU8439800
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only