missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trifluorobenzaldehyde, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 312710010

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC312710010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

161793-17-5
160.09
UQEDGFZRPSAHLC-UHFFFAOYSA-N
519226
C1=CC(=C(C(=C1C=O)F)F)F
C7H3F3O
MFCD00061230
2,3,4-trifluorobenz-aldehyde, benzaldehyde, 2,3,4-trifluoro, 2,3,4-trifluoro benzaldehyde, pubchem4255, intermediates-zcf02215, acmc-209dn7, 2,3,4-trilfuorobenzaldehyde, ksc182o7d, 2,3,4-trifluoro-benzaldehyde, timtec-bb sbb003969
2,3,4-trifluorobenzaldehyde

Specifications

2, 3, 4-Trifluorobenzaldehyde
97.5
1.4040g/mL
56°C
97.5% min. (GC)
C7H3F3O
F3C6H2CHO
MFCD00061230
2,3,4-trifluorobenz-aldehyde, benzaldehyde, 2,3,4-trifluoro, 2,3,4-trifluoro benzaldehyde, pubchem4255, intermediates-zcf02215, acmc-209dn7, 2,3,4-trilfuorobenzaldehyde, ksc182o7d, 2,3,4-trifluoro-benzaldehyde, timtec-bb sbb003969
C1=CC(=C(C(=C1C=O)F)F)F
160.09
160.09
161793-17-5
100.0
77°C to 79°C (20.0mmHg)
Authentic
Glass bottle
1.4800 to 1.482
1g
1.404
UQEDGFZRPSAHLC-UHFFFAOYSA-N
2,3,4-trifluorobenzaldehyde
519226
98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions