missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,6-Tetra-O-benzoyl-D-glucopyranose 90.0+%, TCI America™

Catalog No. T2020250MG
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2020250MG 250 mg
T20201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T2020250MG Supplier TCI America Supplier No. T2020250MG
Only null left

Chemical Identifiers

CAS 64768-20-3
Molecular Formula C34H28O10
Molecular Weight (g/mol) 596.588
MDL Number MFCD00066004
InChI Key FCDYAJBVISGNLC-UCDCFHRCSA-N
PubChem CID 22215583
IUPAC Name [(2R,3R,4S,5R,6R)-3,4,5-tribenzoyloxy-6-hydroxyoxan-2-yl]methyl benzoate
SMILES C1=CC=C(C=C1)C(=O)OCC2C(C(C(C(O2)O)OC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4)OC(=O)C5=CC=CC=C5

Specifications

Melting Point 122°C
Color White-Yellow
Quantity 250 mg
Formula Weight 596.59
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4,6-Tetra-O-benzoyl-D-glucopyranose