missing translation for 'onlineSavingsMsg'
Learn More

2,3:4,6-Di-O-isopropylidene-alpha-L-sorbofuranose, 98%, Thermo Scientific Chemicals

Catalog No. AAJ6619003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6619003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6619003 Supplier Thermo Scientific Chemicals Supplier No. J6619003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17682-70-1 | Molecular Formula: C12H20O6 | Molecular Weight: 260.286 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17682-70-1
Molecular Formula C12H20O6
Molecular Weight (g/mol) 260.286
MDL Number MFCD00132929
InChI Key GQXSDDHYUVYJCQ-HNBLOZHYSA-N
PubChem CID 92220487
IUPAC Name [(3aS,4aR,8aR,8bR)-2,2,7,7-tetramethyl-4a,5,8a,8b-tetrahydro-[1,3]dioxolo[3,4]furo[1,3-d][1,3]dioxin-3a-yl]methanol
SMILES CC1(OCC2C(O1)C3C(O2)(OC(O3)(C)C)CO)C

Specifications

Melting Point 76°C to 80°C
Color White
Quantity 1 g
Formula Weight 260.28
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,3:4,6-Di-O-isopropylidene-alpha-L-sorbofuranose
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only