missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5-Tetrafluorobenzoyl Chloride 97.0+%, TCI America™

Catalog No. T18555G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18555G 5 g
T185525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T18555G Supplier TCI America Supplier No. T18555G
Only null left

Chemical Identifiers

CAS 94695-48-4
Molecular Formula C7HClF4O
Molecular Weight (g/mol) 212.53
MDL Number MFCD00075164
InChI Key XWCKIXLTBNGIHV-UHFFFAOYSA-N
Synonym 2,3,4,5-tetrafluorobenzoylchloride, tetrafluorobenzoyl chloride, 2,3,4,5-tetrafluoro benzoyl chloride, benzoyl chloride, 2,3,4,5-tetrafluoro, 2,3,4,5-tetrafluorbenzoylchlorid, 2,3,4,5-tetrafluoro-benzoyl chloride, 2,3,4,5-tetrafluorobenzoic acid chloride, 2,3,4,5-tetrafluorobezoyl chloride, benzoyl chloride, 2,3,4,5-tetrafluoro-9ci, 2,3,4,5-tetrafluorobezoylchloride
PubChem CID 2733689
IUPAC Name 2,3,4,5-tetrafluorobenzoyl chloride
SMILES FC1=CC(C(Cl)=O)=C(F)C(F)=C1F

Specifications

Color Yellow
Quantity 5 g
UN Number 3265
Formula Weight 212.53
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,3,4,5-Tetrafluorobenzoyl Chloride