missing translation for 'onlineSavingsMsg'
Learn More

2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose 98.0+%, TCI America™

Catalog No. D37585G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37585G 5 g
D375825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37585G Supplier TCI America Supplier No. D37585G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 20880-92-6
Molecular Formula C12H20O6
Molecular Weight (g/mol) 260.286
MDL Number MFCD00022183
InChI Key PSSHGMIAIUYOJF-OZFQHSNDSA-N
Synonym 2,3:4,5-bis-o-1,2 methylethylidene-beta-d-fruetopyranose
PubChem CID 45357248
IUPAC Name [(3aR,5aS,8aS)-2,2,7,7-tetramethyl-5,5a,8a,8b-tetrahydrodi[1,3]dioxolo[4,5-a
SMILES CC1(OC2COC3(C(C2O1)OC(O3)(C)C)CO)C

Specifications

Melting Point 96°C
Color White
Quantity 5 g
Formula Weight 260.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose
TSCA No
Recommended Storage Freezer