Learn More
Alkali Scientific CellPro™ 2.2ml 96-Well Deep Well Plate, V-Bottom, Square Well, Sterile, 5/pk, 50/cs
Numéro de catalogue. 502335921
Encompass
Our deep well plates are compatible with all popular automated research platforms and robotic equipment. The plates are useful for bacterial cultures, centrifugation (up to 4000 x g), and general sample storage. All plates are produced in a class 100,000 cleanroom facility and are ANSI/SLAS 1-2004 compliant. They can be used with silicone sealing mats and sealing films of various materials (sold separately).
Numéro de catalogue. 502335921 Fournisseur Alkali Scientific Code fournisseur MPD26S
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Our deep well plates are compatible with all popular automated research platforms and robotic equipment. The plates are useful for bacterial cultures, centrifugation (up to 4000 x g), and general sample storage. All plates are produced in a class 100,000 cleanroom facility and are ANSI/SLAS 1-2004 compliant. They can be used with silicone sealing mats and sealing films of various materials (sold separately).