missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Thiodiethylene Bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate] 98.0+%, TCI America™

Catalog No. T1070500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1070500G 500 g
T107025G 25 g
T1070100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T1070500G Supplier TCI America Supplier No. T1070500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 41484-35-9
Molecular Formula C38H58O6S
Molecular Weight (g/mol) 642.94
MDL Number MFCD00059346
InChI Key VFBJXXJYHWLXRM-UHFFFAOYSA-N
PubChem CID 64883
IUPAC Name 2-[(2-{[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoyl]oxy}ethyl)sulfanyl]ethyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate
SMILES CC(C)(C)C1=CC(CCC(=O)OCCSCCOC(=O)CCC2=CC(=C(O)C(=C2)C(C)(C)C)C(C)(C)C)=CC(=C1O)C(C)(C)C

Specifications

Melting Point 78°C
Color White
Quantity 500 g
Formula Weight 642.94
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Thiodiethylene Bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate]
RTECSNumber DA8342500
TSCA Yes