missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Thienyl)ethyl p-Toluenesulfonate 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 32°C |
| Quantity | 5 g |
| Formula Weight | 282.37 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-(2-Thienyl)ethyl p-Toluenesulfonate |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |