missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Pyridyl)benzo[b]thiophene 97.0+%, TCI America™

Numéro de catalogue. B35611G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B35611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. B35611G Fournisseur TCI America Code fournisseur B35611G
Il en reste null

Identifiants chimiques

CAS 38210-35-4
Molecular Formula C13H9NS
Molecular Weight (g/mol) 211.282
MDL Number MFCD08457652
InChI Key NRSBAUDUBWMTGL-UHFFFAOYSA-N
Synonym 2-[Benzo[b]thiophen-2-yl]pyridine
PubChem CID 11505853
IUPAC Name 2-(1-benzothiophen-2-yl)pyridine
SMILES C1=CC=C2C(=C1)C=C(S2)C3=CC=CC=N3

Spécifications

Melting Point 125°C
Color White-Yellow
Quantity 1 g
Formula Weight 211.28
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2-Pyridyl)benzo[b]thiophene