missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Propynyloxy)naphthalene 98.0+%, TCI America™
$221.83 - $738.16
Chemical Identifiers
| CAS | 20009-28-3 |
|---|---|
| Molecular Formula | C13H10O |
| Molecular Weight (g/mol) | 182.22 |
| MDL Number | MFCD04275627 |
| InChI Key | RACCOFGYHHVUQB-UHFFFAOYSA-N |
| Synonym | 2-(Propargyloxy)naphthalene |
| PubChem CID | 15689008 |
| IUPAC Name | 2-(prop-2-yn-1-yloxy)naphthalene |
| SMILES | C#CCOC1=CC=C2C=CC=CC2=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P2190200MG
|
TCI America
P2190200MG |
200 mg |
Each for $221.83
|
|
|||||
|
P21901G
|
TCI America
P21901G |
1 g |
Each for $738.16
|
|
|||||
Chemical Identifiers
| 20009-28-3 | |
| 182.22 | |
| RACCOFGYHHVUQB-UHFFFAOYSA-N | |
| 15689008 | |
| C#CCOC1=CC=C2C=CC=CC2=C1 |
| C13H10O | |
| MFCD04275627 | |
| 2-(Propargyloxy)naphthalene | |
| 2-(prop-2-yn-1-yloxy)naphthalene |
Specifications
| 20009-28-3 | |
| White | |
| C13H10O | |
| 200 mg | |
| RACCOFGYHHVUQB-UHFFFAOYSA-N | |
| 2-(prop-2-yn-1-yloxy)naphthalene | |
| 15689008 | |
| ≥98.0% (GC) | |
| 2-(2-Propynyloxy)naphthalene |
| 60°C | |
| 83°C | |
| MFCD04275627 | |
| 2-(Propargyloxy)naphthalene | |
| C#CCOC1=CC=C2C=CC=CC2=C1 | |
| 182.22 | |
| 182.22 | |
| Crystalline Powder |
Safety and Handling
TSCA : No