accessibility menu, dialog, popup

UserName

HCO

CAS RN 20009-28-3

IUPAC Name: 2-(prop-2-yn-1-yloxy)naphthalene
Synonyms: 2-(prop-2-yn-1-yloxy)naphthalene
Molecular Weight (g/mol): 182.22
Molecular Formula: C13H10O
InChi Key: RACCOFGYHHVUQB-UHFFFAOYSA-N
SMILES: C#CCOC1=CC=C2C=CC=CC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)