missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Propynyloxy)naphthalene 98.0+%, TCI America™
Supplier: TCI America P21901G
Specifications
| 2-(2-Propynyloxy)naphthalene | |
| 60°C | |
| 83°C | |
| MFCD04275627 | |
| 2-(Propargyloxy)naphthalene | |
| C#CCOC1=CC=C2C=CC=CC2=C1 | |
| 182.22 | |
| 182.22 | |
| Crystalline Powder |
| 20009-28-3 | |
| White | |
| C13H10O | |
| 1 g | |
| RACCOFGYHHVUQB-UHFFFAOYSA-N | |
| 2-(prop-2-yn-1-yloxy)naphthalene | |
| 15689008 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 20009-28-3 | |
| 182.22 | |
| RACCOFGYHHVUQB-UHFFFAOYSA-N | |
| 15689008 | |
| C#CCOC1=CC=C2C=CC=CC2=C1 |
| C13H10O | |
| MFCD04275627 | |
| 2-(Propargyloxy)naphthalene | |
| 2-(prop-2-yn-1-yloxy)naphthalene |
Safety and Handling
TSCA : No