missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Propynyloxy)naphthalene 98.0+%, TCI America™

Supplier:  TCI America P2190200MG

 View more versions of this product

Catalog No. P2190200MG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-(2-Propynyloxy)naphthalene
60°C
83°C
MFCD04275627
2-(Propargyloxy)naphthalene
C#CCOC1=CC=C2C=CC=CC2=C1
182.22
182.22
Crystalline Powder
20009-28-3
White
C13H10O
200 mg
RACCOFGYHHVUQB-UHFFFAOYSA-N
2-(prop-2-yn-1-yloxy)naphthalene
15689008
≥98.0% (GC)

Chemical Identifiers

20009-28-3
182.22
RACCOFGYHHVUQB-UHFFFAOYSA-N
15689008
C#CCOC1=CC=C2C=CC=CC2=C1
C13H10O
MFCD04275627
2-(Propargyloxy)naphthalene
2-(prop-2-yn-1-yloxy)naphthalene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions