missing translation for 'onlineSavingsMsg'
Learn More

[2.2]Paracyclophane, 99%

Catalog No. AAB2013606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2013606 5 g
AAB2013603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2013606 Supplier Thermo Scientific Chemicals Supplier No. B2013606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1633-22-3 | C16H16 | 208.304 g/mol

2,2]-Paracyclophane is the raw material for the synthesis of parylene which is widely used in microelectronic integrated circuit.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2]-Paracyclophane is the raw material for the synthesis of parylene which is widely used in microelectronic integrated circuit.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 1633-22-3
Molecular Formula C16H16
Molecular Weight (g/mol) 208.304
MDL Number MFCD00003707
InChI Key OOLUVSIJOMLOCB-UHFFFAOYSA-N
Synonym 2.2 paracyclophane, di-p-xylylene, di-1,4-xylylene, paracyclophane, cyclobis benzene-1,4-dimethylene, tricyclo 8,2,2,24,7 hexadeca-4,6,10,12,13,15-hexaene, 2.2-paracyclophane, tricyclo 8.2.2.24,7 hexadeca-4,6,10,12,13,15-hexaene, 2,2-paracyclophane
PubChem CID 74210
SMILES C1CC2=CC=C(CCC3=CC=C1C=C3)C=C2

Specifications

Melting Point 283°C to 288°C
Odor Odorless
Quantity 5 g
Beilstein 1910888
Solubility Information Insoluble in water.
Formula Weight 208.3
Percent Purity 99%
Chemical Name or Material [2.2]Paracyclophane
EINECSNumber 216-644-2
TSCA Yes

RUO – Research Use Only