missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Oxydiacetyl Chloride 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 57°C |
| Quantity | 5 g |
| UN Number | 3265 |
| Formula Weight | 170.97 |
| Percent Purity | ≥97.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 2,2′-Oxydiacetyl Chloride |
Safety and Handling
| EINECSNumber | (2)-3790 |
| TSCA | No |