missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2′-Methylenebis(6-tert-butyl-p-cresol), TCI America™

$27.50 - $73.33

Chemical Identifiers

CAS 119-47-1
Molecular Formula C23H32O2
Molecular Weight (g/mol) 340.507
MDL Number MFCD00043641
InChI Key KGRVJHAUYBGFFP-UHFFFAOYSA-N
Synonym Bis(3-tert-butyl-2-hydroxy-5-methylphenyl)methane, 3,3′C-Di-tert-butyl-2,2′C-dihydroxy-5,5′C-dimethyldiphenylmethane, 2,2′C-Methylenebis(6-tert-butyl-4-methylphenol)
PubChem CID 8398
IUPAC Name 2-tert-butyl-6-[(3-tert-butyl-2-hydroxy-5-methylphenyl)methyl]-4-methylphenol
SMILES CC1=CC(=C(C(=C1)C(C)(C)C)O)CC2=C(C(=CC(=C2)C)C(C)(C)C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M021725G View Documents TCI America
M021725G
25g Each for $27.50
 
M0217250G View Documents TCI America
M0217250G
250g Each for $73.33
 
Specifications

Chemical Identifiers

119-47-1
340.507
KGRVJHAUYBGFFP-UHFFFAOYSA-N
8398
CC1=CC(=C(C(=C1)C(C)(C)C)O)CC2=C(C(=CC(=C2)C)C(C)(C)C)O
C23H32O2
MFCD00043641
Bis(3-tert-butyl-2-hydroxy-5-methylphenyl)methane, 3,3′C-Di-tert-butyl-2,2′C-dihydroxy-5,5′C-dimethyldiphenylmethane, 2,2′C-Methylenebis(6-tert-butyl-4-methylphenol)
2-tert-butyl-6-[(3-tert-butyl-2-hydroxy-5-methylphenyl)methyl]-4-methylphenol

Specifications

White-Yellow
25g
C23H32O2
Bis(3-tert-butyl-2-hydroxy-5-methylphenyl)methane, 3,3′C-Di-tert-butyl-2,2′C-dihydroxy-5,5′C-dimethyldiphenylmethane, 2,2′C-Methylenebis(6-tert-butyl-4-methylphenol)
CC1=CC(=C(C(=C1)C(C)(C)C)O)CC2=C(C(=CC(=C2)C)C(C)(C)C)O
340.507
340.51
≥99.0% (GC)
130°C
119-47-1
MFCD00043641
KGRVJHAUYBGFFP-UHFFFAOYSA-N
2-tert-butyl-6-[(3-tert-butyl-2-hydroxy-5-methylphenyl)methyl]-4-methylphenol
8398
Crystal-Powder at 20°C
2,2′-Methylenebis(6-tert-butyl-p-cresol)
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0100

RTECSNumber : PA3500000

TSCA : Yes